Learn More
Thermo Scientific Chemicals Mithramycin A, 90%, from Streptomyces argillaceus
£111.56 - £1137.40
Chemical Identifiers
CAS | 18378-89-7 |
---|---|
Molecular Formula | C52H76O24 |
Molecular Weight (g/mol) | 1085.156 |
MDL Number | MFCD00135618 |
InChI Key | CFCUWKMKBJTWLW-ZUZMCERRSA-N |
PubChem CID | 122172942 |
IUPAC Name | (2R,3R)-2-[(2R,4S,5S,6S)-4-[(2R,4S,5S,6S)-4-[(2R,4R,5R,6S)-4,5-dihydroxy-4,6-dimethyloxan-2-yl]oxy-5-hydroxy-6-methyloxan-2-yl]oxy-5-hydroxy-6-methyloxan-2-yl]oxy-3-[(1R,3R,4S)-3,4-dihydroxy-1-methoxy-2-oxopentyl]-6-[(2S,4S,5S,6S)-4-[(2S,4S,5R,6S)-4,5-dihShow More |
SMILES | CC1C(C(CC(O1)OC2CC(OC(C2O)C)OC3=CC4=CC5=C(C(=O)C(C(C5)C(C(=O)C(C(C)O)O)OC)OC6CC(C(C(O6)C)O)OC7CC(C(C(O7)C)O)OC8CC(C(C(O8)C)O)(C)O)C(=C4C(=C3C)O)O)O)OShow More |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||||
10599213
|
Thermo Scientific Chemicals
415090010 |
1 mg | Glass Bottle |
£111.56
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
10432971
|
Thermo Scientific Chemicals
415090050 |
5 mg | Glass Bottle |
£368.84
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
10608424
|
Thermo Scientific Chemicals
415090250 |
25 mg | Glass Bottle | N/A | N/A | N/A | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
18378-89-7 | |
1085.156 | |
CFCUWKMKBJTWLW-ZUZMCERRSA-N | |
(2R,3R)-2-[(2R,4S,5S,6S)-4-[(2R,4S,5S,6S)-4-[(2R,4R,5R,6S)-4,5-dihydroxy-4,6-dimethyloxan-2-yl]oxy-5-hydroxy-6-methyloxan-2-yl]oxy-5-hydroxy-6-methyloxan-2-yl]oxy-3-[(1R,3R,4S)-3,4-dihydroxy-1-methoxy-2-oxopentyl]-6-[(2S,4S,5S,6S)-4-[(2S,4S,5R,6S)-4,5-dihShow More |
Specifications
Mithramycin A | |
18378-89-7 | |
100.0 | |
Authentic | |
90% | |
MFCD00135618 | |
Solubility in water: soluble in water. Other solubilities: soluble in lower alcohols, acetone, ethyl acetate, moderatly soluble in chloroform, slightly soluble in ether, benzene | |
CC1C(C(CC(O1)OC2CC(OC(C2O)C)OC3=CC4=CC5=C(C(=O)C(C(C5)C(C(=O)C(C(C)O)O)OC)OC6CC(C(C(O6)C)O)OC7CC(C(C(O7)C)O)OC8CC(C(C(O8)C)O)(C)O)C(=C4C(=C3C)O)O)O)O | |
1085.156 | |
1085.16 | |
Mithramycin A, 90% |
Powder | |
88.0 | |
180.0°C to 183.0°C | |
Brown to Red | |
C52H76O24 | |
15, 7652 | |
CFCUWKMKBJTWLW-ZUZMCERRSA-N | |
(2R,3R)-2-[(2R,4S,5S,6S)-4-[(2R,4S,5S,6S)-4-[(2R,4R,5R,6S)-4,5-dihydroxy-4,6-dimethyloxan-2-yl]oxy-5-hydroxy-6-methyloxan-2-yl]oxy-5-hydroxy-6-methyloxan-2-yl]oxy-3-[(1R,3R,4S)-3,4-dihydroxy-1-methoxy-2-oxopentyl]-6-[(2S,4S,5S,6S)-4-[(2S,4S,5R,6S)-4,5-dih | |
122172942 | |
90% |
Safety and Handling
GHS P Statement
Harmful if swallowed.
Toxic in contact with skin.
GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
missing translation for 'rtecsNumber' : PZ2800000
- Mithramycin A
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
Hazard Statement
- H311-Toxic in contact with skin.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.