missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mitofilin Monoclonal Antibody (MM#7A-2E4AD5), Alexa Fluor™ 555, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA456400A555
This item is not returnable.
View return policy
Description
Mitofilin is a transmembrane protein of the inner mitochondrial membrane and it is associated with a large multimeric protein complex of about 1200 kDa. Mitofilin has critical functions in mitochondrial morphology and mitochondrial fusion and fission, specifically in the formation of tubular cristae and cristae junctions. Abnormal mitochondrial morphology has been implicated in many human diseases such as myopathy, cardiomyopathy, rhabdomyosarcoma and Whartin′s tumor.Specifications
Mitofilin | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4 | |
Q16891 | |
IMMT | |
Human heart mitochondria | |
50 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunofluorescence | |
MMNo.7A-2E4AD5 | |
Alexa Fluor 555 | |
IMMT | |
P89, MINOS2, HMP, PIG4, P87/89, PIG52, P87 | |
Mouse | |
Purified | |
RUO | |
10989 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
For Research Use Only