missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Mitomycin C, contains 2 mg Mitomycin C and 48 mg NaCl
Tienda Thermo Scientific Chemicals Productos
Click to view available options
Quantity:
2 mg
Tamaño de la unidad:
2mg
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General description
- Mitomycin C is a water soluble, blue-violet crystalline powder, unstable in light and acidic conditions, but may be stably stored at 2-8°c
- Bio-reduced mitomycin C generates oxygen radicals, alkylates DNA, and produces inter-strand DNA cross-links, thereby inhibiting DNA synthesis
Applications
- Mitomycin C is preferentially toxic to hypoxic cells that are resistant to cancer therapy, and is therefore used to study its potential therapeutic effect on advanced stages of cancer
- At high concentrations, mitomycin C inhibits RNA and protein synthesis and demonstrates strong antibiotic properties towards both Gram-positive and Gram-negative bacteria
Especificaciones
Especificaciones
| Chemical Name or Material | Mitomycin C |
| CAS | 50-07-7, 7647-14-5 |
| Melting Point | >360.0°C |
| Color | Blue-Violet |
| Packaging | Ampoule |
| Molecular Formula | C15H18N4O5 |
| MDL Number | MFCD00078109 |
| Quantity | 2 mg |
| Merck Index | 15, 6300 |
| Synonym | mitomycin c, mitomycin, ametycine, mutamycin, ametycin, mitocin-c, mitomycinum, mytomycin, mytozytrex, mitomycin-c |
| Mostrar más |
Seguridad y manipulación
Nombre del producto
- Mitomycin C
- Warning
- Acute toxicity Category 4
- Carcinogenicity Category 2
- H351-Suspected of causing cancer.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido