missing translation for 'onlineSavingsMsg'
Learn More

Mitomycin C, Fisher BioReagents™

Product Code. 10115004
Click to view available options
Quantity:
2 mg
10 mg
20 mg
Packaging:
Amber Glass

Mitomycin C

Inhibitor of DNA synthesis, nuclear division, and cancer cells. Antibacterial spectrum: Gram positive, gram negative, acid-fast bacilli.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-07-7
Molecular Formula C15H18N4O5
Molecular Weight (g/mol) 334.332
InChI Key NWIBSHFKIJFRCO-WUDYKRTCSA-N
Synonym mitomycin c, mitomycin, ametycine, mutamycin, ametycin, mitocin-c, mitomycinum, mytomycin, mytozytrex, mitomycin-c
PubChem CID 5746
ChEBI CHEBI:27504
SMILES CC1=C(C(=O)C2=C(C1=O)N3CC4C(C3(C2COC(=O)N)OC)N4)N

Specifications

Melting Point 360°C
Color Blue
Assay Percent Range ≥98.0 %
Packaging Amber Glass
Quantity 20 mg
Merck Index 15, 6300
Solubility Information Soluble in water
Formula Weight 334.3
Percent Purity 99.0% min. (RP CH3CN:H20 0 to 100%), 98.0% min. (RP MEOH:H20 90:10)
Physical Form Solid
Chemical Name or Material Mitomycin C
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Mitomycin C
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'