missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mitomycin C, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
Description
Mitomycin C is an antibiotic and antitumor agent. Covalently binds DNA forming intra- and interstrand crosslinks. Inhibits DNA synthesis. Also used for MEF/STO feeder layer preparation in stem cell culture.
Specifications
Type | Mitomycin C |
CAS | 50-07-7 |
Molecular Formula | C15H18N4O5 |
Synonym | [1aS-(1aα,8β,8aα,8bα)]-6-Amino-8-[[(aminocarbonyl)oxy]methyl]-1,1a,2,8,8a,8b-hexahydro-8a-methoxy-5-methylazirino[2',3':3,4]pyrrolo[1,2-a]indole-4, |
InChI Key | NWIBSHFKIJFRCO-WUDYKRTCSA-N |
SMILES | CC1=C(C(=O)C2=C(C1=O)N3CC4C(C3(C2COC(=O)N)OC)N4)N |
Molecular Weight (g/mol) | 334.332 |
PubChem CID | 5746 |
ChEBI | CHEBI:27504 |
Formula Weight | 334.33 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'