missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ MitoProbe™ Transition Pore Assay Kit, for flow cytometry
MitoProbe™ Transition Pore Assay Kit, for flow cytometry
Brand: Molecular Probes™ M34153
Code : 50
Additional Details : Weight : 0.00450kg
Description
The MitoProbe™ Transition Pore Assay Kit provides a proven method (Petronilli V. et al. 1989, 1999) of measuring mitochondrial permeability transition pore opening. The kit includes a dye and a quencher that are easily loaded into cells. In healthy cells, the dye, but not the quencher, enters mitochondria. The mitochondria remain brightly fluorescent until mitochondrial pore activation permits quenching of the fluorescence.View a selection guide for all apoptosis assays for flow cytometry.
Specifications
100 | |
Transition Pore Assay Kit | |
488 | |
MitoProbe™ |
Tube | |
Contains 5 vials of calcein, AM (50 μg of dry powder/vial), 1 vial of cobalt (II) Chloride hexahydrate (1.2 mL, 80 mM in saline), 1 vial of ionomycin (55 μg), calcium salt, and 1 vial of DMSO (1.5 mL). Store in freezer (-5 to -30°C). |
|
Room Temperature | |
Calcein AM |
For Research Use Only. Not for use in diagnostic procedures.