missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MitoPY1, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
Description
Fluorescent probe for imaging hydrogen peroxide (H2O2) in mitochondria of living cells. Displays enhanced emission at 528 nm in the presence of H2O2. Selective for H2O2 over superoxide, nitric oxide and hydroxyl radical. Detects local increases in H2O2 in an in vitro model of Parkinson's Disease.
Specifications
Specifications
CAS | 1041634-69-8 |
Molecular Formula | C52H53BIN2O5P |
Quantity | 10 mg |
Synonym | mitopy1, 4-4-3-oxo-6'-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl spiro isobenzofuran-1 3h ,9'-9h xanthen-3'-yl-1-piperazinyl butyl triphenyl-phosphonium iodide |
InChI Key | JPCCULDYUUMFAD-UHFFFAOYSA-M |
SMILES | B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4(C5=C(O3)C=C(C=C5)N6CCN(CC6)CCCC[P+](C7=CC=CC=C7)(C8=CC=CC=C8)C9=CC=CC=C9)C1=CC=CC=C1C(=O)O4.[I-] |
IUPAC Name | 4-[4-[3-oxo-6'-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)spiro[2-benzofuran-1,9'-xanthene]-3'-yl]piperazin-1-yl]butyl-triphenylphosphanium;iodide |
Molecular Weight (g/mol) | 954.693 |
PubChem CID | 90488942 |
Formula Weight | 954.68 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'