missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MitoTracker™ Dyes for Mitochondria Labeling

These mitochondria dyes are specially packaged in vials of 50 μg each to create freshly reconstituted dyes without the impact of freeze-thaw cycles to aid in a simplified sample prep for cell analysis workflow.

Brand:  Invitrogen™ M22426

269.04 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 12010156

  • £354.00 / 1000µg

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

Rosamine & carbocyanine-based staining dyes MitoTracker Orange CMTMRos, Red CMXRos, Orange CM-H2TMRos, Red CM-H2XRos, Red FM, Green FM, & Deep Red FM enable mitochondria visualization with fluorescent imaging. These mitochondria dyes are specially packaged in vials of 50 μg each to create freshly reconstituted dyes without the impact of freeze-thaw cycles to aid in a simplified sample prep for cell analysis workflow.

Rosamine-based MitoTracker dyes for mitochondria labeling

Cell-permeant MitoTracker probes stain active mitochondria in live cells for labeling and localization in fluorescent cell imaging. Our rosamine-based mitochondrial staining dyes include MitoTracker Orange CMTMRos, a derivative of tetramethylrosamine, and MitoTracker Red CMXRos, a derivative of X-rosamine. MitoTracker Orange CM-H2TMRos and MitoTracker Red CM-H2XRos are reduced, nonfluorescent versions of the rosamine-based MitoTracker dyes that fluoresce upon oxidation. The fluorescent signal from the rosamine-based MitoTracker dyes is retained in the mitochondria even after aldehyde fixation and detergent permeabilization, making these mitochondria dyes flexible for many workflows including applications that require subsequent processing such as immunocytochemistry or in situ hybridization. MitoTrackerRed and Orange are well suited for multicolor labeling experiments because their fluorescence is well resolved from the green fluorescence of other probes.

Carbocyanine-based MitoTracker dyes for mitochondria labeling

Cell-permeant MitoTracker dyes stain active mitochondria in live cells for mitochondrial labeling and localization in fluorescent cell imaging. The carbocyanine-based dyes MitoTracker Green FM and MitoTracker Red FM accumulate in active mitochondria but are not well-retained in mitochondria after aldehyde fixation. The carbocyanine-based MitoTracker Deep Red FM stains active mitochondrial in live cells and is well-retained in mitochondria after aldehyde fixation and subsequent permeabilization with detergents for applications that require subsequent processing such as immunocytochemistry or in situ hybridization. MitoTracker Red FM and MitoTracker Deep Red FM are well suited for multicolor labeling experiments because their red fluorescence is well resolved from the green fluorescence of other probes.

Benefits of using MitoTracker mitochondria labeling dyes

MitoTracker dyes are provided in vials of 50 μg of lyophilized powder ready for reconstitution. To label mitochondria, live cells are simply incubated with the MitoTracker probe of your choice. The mitochondrial staining dyes passively diffuse across the plasma membrane and accumulate in active mitochondria. The MitoTracker dyes are offered in a range of wavelengths and can be used for mitochondrial localization in multicolor experiments.

Conventional fluorescent stains such as tetramethylrosamine and rhodamine 123 are readily sequestered by active mitochondria and are reversible in dynamic membrane potential measurements as they easily wash out of cells upon loss in membrane potential. In contrast, MitoTracker dyes contain a mildly thiol-reactive chloromethyl moiety so that mitochondrial staining is retained if the mitochondrial membrane potential is lost, allowing many of the MitoTracker dyes to be retained during cell fixation.

Throughout the cell life cycle, mitochondria use oxidizable substrates to produce an electrochemical proton gradient across the mitochondrial membrane (whose potential is negative), resulting in ATP production. MitoTracker dyes are ideal probes for mitochondria staining in experiments studying the cell cycle or processes such as apoptosis and other end point assays. MitoTracker dyes are also available for flow cytometry applications (Cat. No. M46750, M46751, M46752, and M46753).

Related products for mitochondrial membrane potential

For studying dynamic mitochondria membrane potential specifically, we recommend using JC-1 (cationic carbocyanine dye, Cat. No. T3168) or TMRM (tetramethyl rhodamine methyl ester, Cat. No. I34361) dyes. The MitoProbe JC-1 Assay Kit (Cat. No. M34152) contains the JC-1 dye in addition to the potent mitochondrial membrane-potential disrupter, CCCP, which depolarizes the mitochondrial membrane. These reagents can provide compensatory controls to correctly compensate green-to-red fluorescence ratio. TMRM is used for the detection of dynamic measurement of mitochondrial membrane potential.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Deep Red
644/665 nm
Fluorescence Microscope, Flow Cytometer, Microplate Reader
Fluorescence
MitoTracker™
Fluorescent Dye
Mitochondria/Organelle Stain
Store in freezer -5°C to -30°C and protect from light.
Special packaging
20 x 50 μg
MitoTracker™ Deep Red FM
Room Temperature
Dye
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.