Learn More
Invitrogen™ MitoTracker™ Red FM Dye, for flow cytometry
Mitotracker™ dyes for flow cytometry are cell permeant reagents that label mitochondria in live cells. Provided at an optimized concentration with protocols for flow cytometry to allow for more precise analysis with both conventional and spectral flow cytometry.
Brand: Invitrogen™ M46751
Additional Details : Weight : 0.01400kg
Description
- Cell permeant reagents that label mitochondria in live cells
- Dyes are provided at an optimized concentration with protocols for flow cytometry to allows for more precise analysis with both conventional and spectral flow cytometry
- Simply incubate cells in the recommended dye concentration for flow cytometry, which passively diffuses across the plasma membrane and accumulates in active mitochondria
- Bright colors allow for easy detection of mitochondria using flow cytometry without the need for special filters
- Dyes are consistent, reliable, and stable allowing for the generation of high-quality data
- Optimized for flow cytometry use with all essential reagents for labeling mitochondria in a single kit
- Dissolve a vial of dye in anhydrous DMSO to provide enough dye for 20 assays
- A range of available colors are detected with standard flow cytometry filters
Specifications
5 x 20 tests | |
RUO | |
Flow Cytometry | |
5 vials |
100 tests | |
Room Temperature | |
Dye |