Learn More
Micronova™ Mitt wipe controlled environment NovaPoly double thin sponge 305mm PolyMitt
MittWipes™ Wipers offer a high-quality wiper with the security of a glove. The basic mitt fits over a gloved hand, allowing easier cleaning of hard to reach areas around carts, shelving, tanks and tubing. Can also be used as a protective layer over standard gloves.
Brand: Micronova™ MWDT-611
Code : 03
Additional Details : Weight : 1.00000kg
Description
- Microfibre
- Double thin foam interior for increased absorbency
- Fits comfortably over a gloved hand
- Can be used as a protective layer over standard gloves in some clean build and maintenance applications
Specifications
Polyester | |
White | |
Fits comfortably over a gloved hand, for easy cleaning of hard-to-reach areas around carts, shelving, tanks and tubing. Can also be used as a protective layer over standard gloves | |
Foam interior |