missing translation for 'onlineSavingsMsg'
Learn More

Long List Appendix IX Mix E, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Brand:  SPEX Certiprep 8260-E

Code : NEW

Additional Details : CAS Number : 67-56-1 Weight : 0.00100kg Transport : UN number : 1230 Chem class : 3 Pack group : II


Product Code. 15285846

  • £37.60 / 1mL
Estimated Shipment: 11-10-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
2-Butanone78-93-3200μg/mL
2-Chloroethyl vinyl ether110-75-8200μg/mL
2-Hexanone591-78-6200μg/mL
4-Methyl-2-pentanone108-10-1200μg/mL
Acetone67-64-1200μg/mL
Carbon disulfide75-15-0200μg/mL
Vinyl acetate108-05-4200μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Long List Appendix IX Mix E
2mL amber prescored glass ampule
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
200μg/mL
Methanol-P&T
Method 8260B
1 mL
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
For US EPA Method 8260B
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Long List Appendix IX Mix E

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : methanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product intended for laboratory use only