Learn More
Macherey-Nagel™ Mix III Diamino Clean-Up Mix
Brand:  Macherey-Nagel™ 730972
Code : 50
Additional Details : Weight : 0.20000kg
Product Code. 12820961
  • £92.95 / Each
Estimated Shipment: 04-10-2024
to see stock.
Add to basket
Specifications
Chromabond QuEChERS Clean-up Mixes (EN 15662)
Diamino Clean-Up Mix
15 mL
50 Pack