missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Terracycle™ Mixed Beard Nets, Earplugs and Hair Nets Zero Waste Box™

Recycle Mixed Beard Nets, Earplugs and Hair Nets using this Zero Waste Box system. Zero Waste Box includes signage, a built-in liner, shipping to and from your location, and processing of the materials.
£117.50 - £214.13
Specifications
Color | Beige, Green, Red, White |
---|---|
Disposable | Single-use |
Latex Free | Yes |
Sterility | Non-sterile |
Autoclavable | Not Autoclavable |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
17855815
![]() |
Terracycle™
CS46-S-UK |
Small |
£117.50
Each |
Estimated Shipment: 24-04-2025
Log in to see stock. |
||||||
17845715
![]() |
Terracycle™
CS46-M-UK |
Medium |
£153.13
Each |
Estimated Shipment: 24-04-2025
Log in to see stock. |
||||||
17825725
![]() |
Terracycle™
CS46-L-UK |
Large |
£214.13
Each |
Estimated Shipment: 24-04-2025
Log in to see stock. |
||||||
Description
Use this Zero Waste Box to recycle:
- Beard nets and hairnets
- Disposable shoe covers
- Earplugs
Do not send in:
- Medical waste
- This includes material contaminated with blood or bodily fluids and waste that originate from health care facilities, hospitals, home care, physicians' offices, dental offices, blood banks, veterinary clinics, research laboratories, etc.
- Disposable masks or gloves
- Lab coats or other garments
- Hazardous waste (sharp, flammable, reactive, corrosive, ignitable, toxic, infectious or pathogenic) which presents a danger to the environment, or to people
- Batteries, pressurized canisters and broken glass
Specifications
Beige, Green, Red, White | |
Yes | |
Not Autoclavable | |
Recycling Box |
Single-use | |
Non-sterile | |
Cardboard, Plastic |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Terracycle™ Mixed Beard Nets, Earplugs and Hair Nets Zero Waste Box™