missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mixed Calibration Standard 1A For Environmental Analysis, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
£345.00
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11415918
|
SPEX Certiprep
MIXSTD1A-500 |
500 mL | Triple-leached LDPE bottle |
£345.00
500mL |
Estimated Shipment: 29-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
As | 100μg/mL |
Ca | 100μg/mL |
Sb | 50μg/mL |
Se | 50μg/mL |
B | 20μg/mL |
Cd | 20μg/mL |
Cu | 20μg/mL |
Mn | 20μg/mL |
Ba | 10μg/mL |
Ag | 5μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Product intended for laboratory use only