Learn More
Mixed Calibration Standard 2 for ICP, 5 Components, Honeywell Fluka™
Brand: Honeywell Fluka E2095-100ML
Additional Details : Weight : 0.10600kg Transport : UN number : 2031 Chem class : 8 Pack group : II
Description
Honeywell Fluka™ analytical standards offer a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. The products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.
Specifications
Mixed Calibration Standard 2 for ICP | |
1.06 g/cm3 | |
100 mL |
Standards for ICP and ICP-MS | |
<2 |