Learn More
Kinematica™ Mixing Attachment for Microtron™ MB 550 Laboratory Mixer
For the mixing, dispersing, homogenizing, emulsifying and size reduction of cuttable materials.
Brand:  Kinematica™ 36050015
Code : A0
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11950049
  • £449.00 / Each
Estimated Shipment: 20-09-2024
to see stock.
Add to basket
Description
Description
  • For use with the MB 550 Mixers
  • The two stainless steel ball bearings are sealed twice each
  • Noiseless and sturdy coupling with dampers
  • Blades, sealing and dampers can easily be exchanged
  • Four high-grade blades designed for optimal mixing
  • Generally suited for intensive and fast processing
Specifications
Specifications
Mixing Head Attachment
Stainless Steel
MB 550 Laboratory Mixer
1 Ea.
500 mL
Borosilicate Glass
Gasket and blade head
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products