missing translation for 'onlineSavingsMsg'
Learn More

Tocris™ ML 335

Selective K2P2.1 (TREK-1) and K2P10.1 (TREK-2) activator

£126.00 - £443.00

Specifications

Molecular Formula C15H14Cl2N2O3S
Formula Weight 373.25
Product Type Activator
Purity 98%
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18727406
View Documents
Tocris™
6887/10
10 mg
£126.00
10mg
Please to purchase this item. Need a web account? Register with us today!
18705796
View Documents Promotion Details
Tocris™
6887/50
50 mg
£443.00
50mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ML 335 is a selective K2P2.1 (TREK-1; KCNK2) and K2P10.1 (TREK-2; KCNK10) activator (EC50 values of 14.3 μM and 5.2 μM, respectively); directly stimulates the C-type gate via the K2P modulator pocket. Does not activate K2P4.1 (TRAAK).
Specifications

Specifications

C15H14Cl2N2O3S
373.25
Activator
98%
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at 4°C

SDS
Documents

Documents

Product Certifications