Learn More
Tocris Bioscience™ ML 354
Selective PAR4 antagonist
Brand:  Tocris Bioscience™ 5387/50
 View more versions of this product
Product Code. 18718198
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
ML 354 is a selective PAR4 antagonist (IC50 = 140 nM). Exhibits 70-fold selectivity for PAR4 over PAR1.
Specifications
Specifications
>98%
282.29
50 mg
ML 354
ML 354