missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ML 354
Selective PAR4 antagonist
£144.70 - £532.38
Specifications
Purity | >98% |
---|---|
Description | ML 354 |
Molecular Weight (g/mol) | 282.29 |
Product Type | ML 354 |
Description
Selective PAR4 antagonist (IC50 = 140 nM). Exhibits 70-fold selectivity for PAR4 over PAR1.Specifications
>98% | |
ML 354 | |
282.29 | |
ML 354 |
For Research Use Only