Learn More
Tocris Bioscience™ ML 365
Potent and selective K2P3.1 (TASK-1) channel blocker
Brand:  Tocris Bioscience™ 5337/10
 View more versions of this product
Product Code. 18736158
  • £182.00 / 10mg
Explore more special offers
Description
Description
ML 365 is a potent and selective TASK-1 (K2P3.1/KCNK3) channel blocker (IC50 values are 4 and 390 nM at TASK-1 and TASK-3, respectively). Displays little or no inhibition at Kir2.1, voltage-gated potassium channels, KCNQ2 and KV11.1 (hERG).
Specifications
Specifications
>99%
ML 365
ML 365
10 mg
360.41