Learn More
Sartorius mLINE™ Pipette Multipacks
Replenish or set up a new pipetting station. Sartorius mLINE™ Pipette Multipacks conveniently combine a set of mLINE mechanical pipettes with matching tip trays and useful accessories. Available in collections of 3, 4 and 5 pipettes with matching tips
Brand:  Sartorius LH-725664
 View more versions of this product
Product Code. 15293583
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Multipacks to match every need
  • Affordable options to get a set of quality pipettes with useful accessories
  • Ideal for renewing an existing pipette collection or setting up a new pipetting station
  • Includes a linear pipette rack, calibration tool, tube opener and color-coding caps
  • Collections include up to five different volumes of single channel pipettes, from 10μL to 5mL; the 3+1 pack includes a 350μL eight-channel pipette
mLINE&trade mechanical pipettes
  • Offer excellent ergonomics, performance, precision and safety in manual pipetting
  • Requires exceptionally light pipetting and tip ejection forces
  • Specifically designed to prevent Repetitive Strain Injury (RSI) in repetitive pipetting
  • Minimized risk of contamination with Safe-Cone Filters and full autoclavability
  • Increased safety with volume lock, preventing accidental volume changes while pipetting
Optifit Tips single trays
  • Designed for the best possible fitting and sealing for the highest possible accuracy and precision
  • Tips are RNase, DNase and endotoxin free and compliant with ISO 9001 and ISO 13485
  • Environment-friendly materials and fully autoclavable to reduce waste
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Combines mLINE pipets and Optifit Tips with for safe, precise pipetting
Repetitive Manual Pipetting
mLINE
Repetitive manual pipetting
5 pipettes with 5 boxes of Optifit Trays
Autoclavable
1
Pipette Package
Pipets: 20μL, 100μL, 200μL, 1000μL and 5000μL, single channel
Optifit Tip tray boxes: 3 x 200μL, 1000μL and 5000μL
Linear stand and accessories
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products