missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMAD-d8, MedChemExpress
MedChemExpress D8-MMAD is a deuterated form of MMAD, which is a microtubule disrupting agent.
Brand: Medchem Express HY-15581S
This item is not returnable.
View return policy
Description
D8-MMAD is a deuterated form of MMAD, which is a microtubule disrupting agent.Specifications
MMAD-d8 | |
Off-White | |
C41H58D8N6O6S | |
DMSO : ≥ 100 mg/mL (128.35 mM) | |
779.11 | |
99.12% | |
Cancer-programmed cell death |
10 mg | |
Research | |
Demethyldolastatin 10-d8 Monomethylauristatin D-d8 Monomethyl Dolastatin 10-d8 | |
O=C(N[C@H](C1=NC=CS1)CC2=CC=CC=C2)[C@H](C)[C@@H](OC)[C@H]3N(C(C[C@@H](OC)[C@H]([C@@H](C)CC)N(C([C@@](C([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])([2H])NC([C@H](C(C)C)NC)=O)=O)C)=O)CCC3 | |
779.11 | |
Research | |
Solid |
Research purposes only