missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ mMESSAGE mMACHINE™ T3 Transcription Kit

Product Code. 10665605
Click to view available options
Product Type:
T3 In Vitro Transcription kit
No. of Reactions:
25 Reactions
Unit Size:
25 reactions
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10665605

Brand: Invitrogen™ AM1348

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Obtain 10–50 times yield of conventional in vitro transcription reactions.

mMESSAGE mMACHINE high yield capped RNA transcription kits use patented high-yield transcription technology for the routine synthesis of 15–35 μg of 7-methyl guanosine capped RNA from 1 μg of template DNA. Each kit includes sufficient reagents for 25 reactions.

  • High yields of capped RNA—up to 35 μg of RNA per reaction
  • Extra convenience—cap analog-nucleotide mix provided
  • Simplified reaction format
  • Synthesized transcripts ideal for microinjection, in vitro translation, or transfection
  • 10–50 times yield obtained with conventional in vitro transcription reactions
  • Ratio of cap analog to GTP has been optimized to allow compromise between yield (15–35 μg) and proportion of transcripts that are capped (80%)
  • Contains a LiCl precipitation solution, efficient for separating proteins and unincorporated nucleotides (including free cap analog) from the capped RNA, allowing an increased efficiency of translation
  • Optimized for use with polymerases included in kit
  • Contains 57 U of cap analog
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage T3 Enzyme Mix, 10X Reaction Buffer 2XNTP/CAP Solution, GTP Solution, pTRI-Xef, TURBO DNase, Ammonium Acetate Stop Solution, Lithium Chloride Precipitation Solution, and Gel Loading Buffer II are all stored at –20°C. Nuclease-free Water may be stored at any temperature.
Final Product Type Synthetic mRNA, mRNA
Format Kit
Sample Type mRNA, Synthetic mRNA
For Use With (Application) In Vitro Transcription, synthetic mRNA, mRNA, transfection, LNPs, GENEart, linearized DNA plasmid, delivery, PCR products
No. of Reactions 25 Reactions
Product Line Ambion, mMESSAGE mMACHINE
Product Type T3 In Vitro Transcription kit
Quantity 25 Reactions
Promoter T3
Shipping Condition Dry Ice
Yield 20-30 μg
Starting Material PCR Products, Linearized DNA plasmid
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.