missing translation for 'onlineSavingsMsg'
Learn More

MNI-caged-L-glutamate, Tocris Bioscience™

Product Code. 18075052
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Stable photoreleaser of L-glutamate

MNI-caged-L-glutamate is a form of glutamate linked to a photo-protecting group, 4-methoxy-7-nitroindolinyl (MNI); it rapidly and efficiently releases L-glutamate (Cat. No. 0218) by photolysis (300 - 380 nm excitation) with a quantum yield in the 0.065-0.085 range. It is also suitable for use with two-photon uncaging microscopy (cross-section of 0.06 GM at 730 nm). MNI-caged-L-glutamate is optically compatible with other chromophores used for fluorescence imaging, such as GFP, YFP and most Ca2+ dyes. MNI-caged-L-glutamate is 2.5-fold more efficient at releasing L-glutamate than NI-caged L-glutamate. MNI-caged-L-glutamate is water-soluble, stable at neutral pH, highly resistant to hydrolysis and pharmacologically inactive at neuronal glutamate receptors and transporters (up to mM concentrations). MNI-caged-L-glutamate can be used for in situ studies of fast synaptic glutamate receptors.

View more information regarding MNI-caged-L-glutamate .

Chemical Identifiers

CAS 295325-62-1
Molecular Formula C14H17N3O6
Molecular Weight (g/mol) 323.305
InChI Key GXIDBZKXGUNITQ-VIFPVBQESA-N
Synonym mni-caged-l-glutamate, 4-methoxy-7-nitroindolinyl-l-glutamate, 2s-2-amino-5-4-methoxy-7-nitro-2,3-dihydroindol-1-yl-5-oxopentanoic acid, mni glutamate, tocris-1490, 4-methoxy-7-nitroindolinyl-caged-l-glutamate, s-2-amino-5-4-methoxy-7-nitroindolin-1-yl-5-oxopentanoic acid, 2s-2-amino-5-4-methoxy-7-nitro-2,3-dihydro-1h-indole-1-yl-5-oxopentanoic acid, s-?-amino-2,3-dihydro-4-methoxy-7-nitro-?-oxo-1h-indole-1-pentanoic acid, s-, a-amino-2,3-dihydro-4-methoxy-7-nitro-d-oxo-1h-indole-1-pentanoic acid
PubChem CID 6604871
IUPAC Name (2S)-2-amino-5-(4-methoxy-7-nitro-2,3-dihydroindol-1-yl)-5-oxopentanoic acid
SMILES COC1=C2CCN(C2=C(C=C1)[N+](=O)[O-])C(=O)CCC(C(=O)O)N

Specifications

Quantity 10 mg
Formula Weight 323.3
Percent Purity >98%
Chemical Name or Material MNI-caged-L-glutamate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.