Learn More
Pal International™ Polypropylene Hairnet Mob Caps
Click to view available options
Size:
One Size
Color:
Blue
Red
White
Description
- Soft, comfortable, thermally-bonded polypropylene fiber construction
- Encapsulated double-elastic edge
- Food safe
- Latex-free to reduce the risk of irritation.
Specifications
Specifications
Product Type | Mob Cap |
Size | One Size |
Material | Polypropylene |
Disposable | Single-use |
Model | D93330HP |
Quantity | 1000/Pk. |
Includes | Encapsulated Double-elastic Edge |