Learn More
Köttermann Mobile Cabinet with One Door
This Mobile Cabinet with One Door offers versatile storage space with a single door to protect stored items.
Brand:  Köttermann 315-00071
 View more versions of this product
Product Code. 17312109
  • £553.00 / Each
Estimated Shipment: 10-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Versatile storage space
  • Mobile with lockable wheels
  • Features one door to protect stored items
  • Many sizes and configurations available
Specifications
Specifications
White
516 x 600 x 740 mm
Epoxy Coated
Right
1
1
Plastic with Brakes
Steel
Mobile
Hinged
Steel
No
None
None
Yes