Learn More
Miele Professional™ Mobile Unit E 385
Product Code. 10129811 Shop All Miele Products
Click to view available options
Includes:
16 Injector Nozzles
Unit Size:
Each
Product Code. 10129811
Brand: Miele Professional™ 3809930
Only null left
Add to basket
Add to basket
Mobile unit E 385 for the optimum loading of narrow necked glassware
  • Equipped with 16 injector nozzles
  • Equipped with connection for hot air drying
  • Bracket for ML/2 for mobile unit recognition / spray arm control
TRUSTED_SUSTAINABILITY
Specifications
Product Type Mobile Injector Unit 1/2
Model E 385
For Use With (Equipment) Narrow-Necked Glassware
Includes 16 Injector Nozzles
Quantity 1 Each