Learn More
Köttermann Mobile Single Bench
This Mobile Single Bench fulfills a variety of tasks in the laboratory.
Brand: Köttermann 505-00171
Additional Details : Weight : 40.31340kg
Description
- Available for sitting and standing work height
- Features castors for mobility
Specifications
750 x 900 x 750 mm | |
Trespa™ TopLab™ PLUS Worktop | |
1 |