missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MOBKL1A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12790-1-AP-20UL
This item is not returnable.
View return policy
Description
MOB4A is similar to the yeast Mob1 protein. Yeast Mob1 binds Mps1p, a protein kinase essential for spindle pole body duplication and mitotic checkpoint regulation. Mob4A regulates NDR1 and NDR2 kinase activity, widely expressed nuclear serine-threonine kinases that have been implicated in cell proliferation and/or tumor progression.Specifications
MOBKL1A | |
Polyclonal | |
Unconjugated | |
MOB1B | |
MATS2, Mob1 homolog 1A, Mob1A, Mob1B, MOB4A, MOBKL1A, Protein Mob4A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
360920, 68473, 92597 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
0.83 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q7L9L4, Q8BPB0 | |
Mob1b | |
MOBKL1A Fusion Protein Ag3509 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction