missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ MOCS3 293T Cell Transient Overexpression Lysate (Denatured) (T01)
Human overexpression lysate
Brand: Abnova™ H00027304-T01.100uL
Additional Details : Weight : 0.00010kg
Specifications
Western Blot | |
MOCS3 293T Cell Transient Overexpression Lysate(Denatured) | |
Theoretical M.W. 50.71kDa | |
Transient overexpression cell lysate was tested with anti-MOCS3 antibody (Mfr. No. H00027304-B01) by Western Blot. | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
293T | |
NM_014484 | |
pCMV-MOCS3 full-length | |
100 μL | |
Human |