missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Modafinil
Psychostimulant
£95.00 - £304.00
Specifications
Chemical Name or Material | 2-[(Diphenylmethyl)sulfinyl]acetamide |
---|---|
CAS | 68693-11-8 |
Target | Additional Adrenergic Receptor-related Compounds |
Molecular Formula | C15H15NO2S |
Purity | 0.98 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18762544
|
Tocris Bioscience™
1811/50 |
50 mg |
£304.00
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18762614
|
Tocris Bioscience™
1811/10 |
10 mg |
£95.00
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Modafinil is a psychostimulant that displays neuroprotective and antiparkinsonian activity in a primate model of Parkinson's disease. Modafinil promotes vigilance and wakefulness without the central and peripheral side effects associated with conventional dopaminergic psychostimulants. Mechanism of action is not fully understood, possibly via modulation of catecholamine/GABAergic neurotransmission.Specifications
2-[(Diphenylmethyl)sulfinyl]acetamide | |
Additional Adrenergic Receptor-related Compounds | |
0.98 |
68693-11-8 | |
C15H15NO2S |
Safety and Handling
Recommended Storage : Store at +4°C
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title