missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tychem 6000 F CHA5 Hooded Coverall

Offering excellent chemical permeation protection to an extensive range of chemicals. DuPont™ Tychem™ 6000 F Hooded Coveralls are made from a lightweight and durable fabric consisting of a proprietary barrier film-laminated to a heavy duty Tyvek substrate.

Brand:  DuPont™ D13395221

593.12 GBP valid until 2024-07-31
Use promo code "21674" to get your promotional price.



Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11793083

  • £773.00 / Pack of 25
Estimated Shipment: 21-08-2024
to see stock.
Add to basket

Description

Description

  • Available in grey for discretionary needs
  • Elasticated face, wrists, waist and ankles for optimal fit
  • Thumb loops prevent sleeves from riding up
  • Self-adhesive chin flap for tight seal of suit to face masks
  • Self-adhesive double zipper flap closure system
  • Stitched and over-taped seams with barrier tape for protection and strength
  • Protects personnel against numerous toxic industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, radioactive contamination, biohazards and certain chemical warfare agents
  • Used for a broad range of applications from chemical spill clean-up, emergency response to military and petrochemical applications
Specifications

Specifications

Hooded Coveralls
Tychem
Elastic
Elastic
Elastic
Stitched, Taped
Chemicals
(EU) 2016/425, EN 14126, EN 1073-2, EN 1149-5
For a broad range of applications from chemical spill clean-up, emergency response, to military and petrochemical applications
Large
Gray
Elastic
Hooded
Double Zipper with Self-adhesive Flap
Category III, Type 3-B, 4-B, 5-B and 6-B
Self-adhesive chin flap
100 to 108 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers