missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyve 600 Plus Green Hooded Coverall
733.76 GBP valid until 2024-07-31
Use promo code "21674" to get your promotional price.
Use promo code "21674" to get your promotional price.
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Description
- Self-adhesive chin strap for tight seal of suit to face mask
- Stitched and over-taped seams for protection and strength
- Tyvek zipper and zipper flap for enhanced protection
- Elasticated face, wrists and ankles as well as a glued-in elastic waist
- Elasticated thumb loops prevent sleeves from riding up
- Permeable to both air and water vapour, yet repels water-based liquids and aerosols
- Excellent barrier against fine particles and fibres (down to 1 micron in size), is ultra-low-linting and antistatically treated
- Applications include maintenance and dismantling jobs in the nuclear industry, pharmaceutical manufacturing or research, biosecurity laboratories as well as in medical applications and when exposed to biological hazards
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Product Type | Hooded Coveralls |
Size | Medium |
Material | Tyvek |
Color | Green |
Waist Style | Elastic |
Ankle Style | Elastic |
Cuff Style | Elastic |
Neck Style | Hooded |
Hood Type | Elastic |
Closure Type | Zipper with Storm Flap |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'