Learn More
3M™ Modul-R™ Half Visor 71361-00003
Product Code. 15779677
Click to view available options
Lens Tint: Clear
3M™ Modul-R™ Half Visor 71361-00003 provides complete face protection against power-washing, painting, chemicals, and heavy mechanical work.
  • Half visor with adjustable ventilation system (close or open position)
  • Can only be used in conjunction with the 3M Modul-R goggle
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 166: 2001
Description Ventilated Half Visor
Lens Tint Clear
For Use With (Equipment) Modul-R™ goggles
For Use With (Application) Construction
Lens Material Polycarbonate