Learn More
Elma™ Modular Baskets
Modular basket system for the Elmasonic P, Elmasonic Select, and EASY.
£11.80 - £34.80
Specifications
Type Small Basket
For Use With (Equipment) Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Select, Elmasonic EASY
View More Specs
Products 5
Product Code Brand Diameter (Metric) Height (Metric) Style Price Quantity & Availability  
Product Code Brand Diameter (Metric) Height (Metric) Style Price Quantity & Availability  
17851337
View Documents
Elma™
3000006017
21 mm 12 mm 4 Sections
£11.80
Each
Estimated Shipment: 17-04-2025
to see stock.
Add to basket
17841337
View Documents
Elma™
3000006018
64 mm 2 mm 9 Sections
£11.80
Each
Estimated Shipment: 17-04-2025
to see stock.
Add to basket
17821337
View Documents
Elma™
3000006012
64 mm 16 mm 1 Section
£18.30
Each
Estimated Shipment: 17-04-2025
to see stock.
Add to basket
17821347
View Documents
Elma™
3000006013
64 mm 16 mm 5 Sections
£30.45
Each
Estimated Shipment: 17-04-2025
to see stock.
Add to basket
17811347
missing translation for 'viewDocuments'
Elma™
3000009104
64 mm 42 mm 1 Section
£34.80
Each
Estimated Shipment: 17-04-2025
to see stock.
Add to basket
Description
Description
  • Stainless steel with plastic handles (10 to 120 and 180 to 300) or Rilsan coating (150)
  • Ergonomic handles
  • Autoclavable
  • Bayonet holes in the bottom for mounting holders (e.g. Elma Pin, silicone holder bottom comb)
  • Square openings in the side walls for mounting silicone holders
Specifications
Specifications
Small Basket
Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Select, Elmasonic EASY
Videos
SDS
Documents
Documents
Certificates
Product Content Correction
Product Title
Elma™ Modular Baskets