missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ Modular Blocks and Accessories

Product Code. 15732569 Shop All Ohaus Products
Click to view available options
Sample Type:
1.5mL Tube
10mm Tube
15/16mm Tube
15mL Tube
50mL Tube
Type:
Dry Block
Temperature Probe
Depth (Metric) Well:
3.91 cm
4.45 cm
4.76 cm
4.8 cm
Diameter (Metric) Well:
1.07 cm
1.1 cm
1.71 cm
1.75 cm
2.9 cm
Unit Size:
Each
This item is not returnable. View return policy
To avoid possible electrical hazard, DO NOT fill well or block with water or other liquids. Units are designed as a dry bath/incubator.

Product Code. 15732569

Brand: OHAUS™ 30400151

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
To avoid possible electrical hazard, DO NOT fill well or block with water or other liquids. Units are designed as a dry bath/incubator.

Constructed from solid anodized aluminum block.

Modular blocks are sold separately from heaters.

  • Close contact of tubes-to-block-walls allow for maximum heat retention
  • Each block has a thermometer well for measuring block temperature
TRUSTED_SUSTAINABILITY

Specifications

Description Module Block 10 mm Test Tube
For Use With (Equipment) Dry Block Heater
Material Anodized Aluminum
No. of Places 24
Type Dry Block
Sample Type 10mm Tube
Length (English) 3.75 in.
Length (Metric) 9.5 cm
Height (English) 2 in.
Height (Metric) 5.1 cm
Depth (English) Well 1.9 in.
Depth (Metric) Well 4.8 cm
Diameter (English) Well 0.42 in.
Diameter (Metric) Well 1.07 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.