Learn More
OHAUS™ Modular Blocks and Accessories
Constructed from solid anodized aluminum block.
Brand: OHAUS™ 30400172
Additional Details : Weight : 1.00000kg
Alert:
To avoid possible electrical hazard, DO NOT fill well or block with water or other liquids. Units are designed as a dry bath/incubator.
Description
Modular blocks are sold separately from heaters.
- Close contact of tubes-to-block-walls allow for maximum heat retention
- Each block has a thermometer well for measuring block temperature
Specifications
Module Block 15 mL Conical Centrifuge | |
Anodized Aluminum | |
Block | |
1 Block | |
9.5 cm | |
5.1 cm | |
4.45 cm | |
1.71 cm |
Dry block heater | |
12 | |
15mL Tube | |
3.75 in. | |
2 in. | |
1.75 in. | |
0.67 in. |