Learn More
OHAUS™ Modular Blocks and Accessories
Constructed from solid anodized aluminum block.
Brand: OHAUS™ 30400246
Additional Details : Weight : 1.10000kg
Alert:
To avoid possible electrical hazard, DO NOT fill well or block with water or other liquids. Units are designed as a dry bath/incubator.
Description
Features an LED display on front panel.
- Enables unit to read actual block
- Includes SS RTD probe, 45.7cm stainless steel support rod, thermometer/temperature probe extension clamp and hook connector
Specifications
Stainless Steel Probe Kit | |
Temperature Probe Kit | |
— | |
— |
Dry block heater | |
Stainless Steel RTD probe, 18 in. (45.7cm) vertical support rod, thermometer/probe extension clamp and hook connector | |
— |