Learn More
Chromatography Research Supplies Large-Capacity Moisture Trap without Base Plate
The Large-Capacity Moisture Trap without Base Plate offers quick and easy install and removal without disconnecting fittings from the line.
Brand: Chromatography Research Supplies 34651-20
Code : NEW
Additional Details : Weight : 0.50000kg
Description
- Suitable for drying laboratory air or central air supplies.
- 1/8 in. brass fittings.
- Traps remove moisture, oils, and dust from carrier gases.
- Large 400-mL capacity means fewer refillings are required.
- Adsorbent materials are indicating desiccant and 5-Å molecular sieve.
- Desiccant changes color from blue to pink when depleted.
- The silicone O-ring seals maintain pressures to 100 psi.
- Traps feature transparent acrylic housing.
- Measure 17 in. L x 2 in. dia.
- Choose a trap with base plate for benchtop mounting.
- Shipping weight 3 lb. (1.4 kg).
Specifications
5 A | |
Suitable for drying laboratory air or central air supplies. | |
Graphite/Plastic | |
13.5 oz. | |
Moisture Trap |
2 in. | |
431.8 mm | |
100 psi | |
400 mL | |
50.8 mm |
Safety and Handling
- CRS SPure H2O Filters
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 1A
Hazard Statement
- H350-May cause cancer.
Precautionary Statement
- P102-Keep out of reach of children.
- P103-Read label before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : quartz