missing translation for 'onlineSavingsMsg'
Learn More

Molecular Biological Water, Waternation

Product Code. 30130303
Click to view available options
Quantity:
10 x 100 mL
10 x 500 mL
6 x 1000 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Molecular Biological Water is nuclease free, endotoxin and particle free water. Complies with the requirements of ASTM D1193-06.

  • Ready to use
  • No chemical additives
  • Batch traceability
  • Certificate of conformity and analysis
  • Reduce expensive test fails due to impurities
  • High performance
  • High accuracy
  • Suppress water-forming side reactions
  • Provide consistent performance across many sample types
The MLG range, a comprehensive suite of five high grade pure waters, in assorted sizes, ranging from simple deionised to molecular biology grades, catering to the majority of pure water lab uses.

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

pH 7
Conductivity 0.1 μS/cm
Grade Molecular Biology
Packaging PET Plastic Bottle
Certificate of Analysis Yes
Quantity 10 x 100 mL
For Use With (Application) Antibody Testing
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'