missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Molecular Biology Reagents
BIOPHARMA

Used in molecular biology, nucleic acid purification applications

Brand:  Corning™ 46-011-CM

 View more versions of this product

Product Code. 15353711

  • £279.67 / Pack of 6

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Certificate of Analysis available
  • Product can be diluted to obtain the desired concentration

Order Info

Shipped at ambient temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Tris Borate EDTA Buffer
8.4 ±0.1
Sterile
10X
6 x 1 L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number02721007 Certificate TypeCertificate of Analysis Date17/12/2021 Catalog Number46-011-CM
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ Molecular Biology Reagents > TBE Buffer; 10X; 6 x 1L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.