missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Molecular Biology Reagents

Used in molecular biology, nucleic acid purification applications

Brand:  Corning™ 46-030-CM

Code : NEW

Additional Details : Weight : 6.98000kg

 View more versions of this product

Product Code. 15383711

  • £314.92 / Pack of 6
Estimated Shipment: 26-11-2024
to see stock.

Description

Description

  • Certificate of Analysis available
  • Product can be diluted to obtain the desired concentration

Order Info

Shipped at ambient temperature

Specifications

Specifications

Tris Hydrochloride Buffer
7.5 ±0.1
Sterile
1M Tris-Hydrochloride Buffers, 1M Tris-HCI; 6 x 1L
6 x 1 L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers