Learn More
Thermo Scientific™ SoftFit-L™ Non-Filtered Low Retention Pipette Tip in Reload Tower
GREENER_CHOICE
Pipette samples with the accuracy and precision you expect from your pipette every time, with these low retention pipette tips that deliver maximum volume of sample.
£44.95 - £71.86
Specifications
Filtered Non-filtered
Color Natural
Clean Claims DNase, RNase, DNA, ATP, and Endotoxin Free
Format Refill
Tip Style Universal
View More Specs
Products 9
Product Code Brand Volume (Metric) Sterility Price Quantity & Availability  
Product Code Brand Volume (Metric) Sterility Price Quantity & Availability  
11575474
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
3722-05-RI
20 μL Non-sterile
£50.95
Pack of 960
Estimated Shipment: 16-07-2025
to see stock.
Add to basket
11585474
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
3752-05-RI
200 μL Non-sterile
£51.10
Pack of 960
In Stock
to see stock.
Add to basket
11515484
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
3732-05-RI
300 μL Non-sterile
£46.20
Pack of 960
Estimated Shipment: 12-08-2025
to see stock.
Add to basket
11535484
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
3782-05-RI
1000 μL Non-sterile
£44.95
Pack of 768
Estimated Shipment: 06-08-2025
to see stock.
Add to basket
11555484
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
3742-05-RI
1200 μL Non-sterile
£64.50
Pack of 768
Estimated Shipment: 17-07-2025
to see stock.
Add to basket
11595494
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
3721-05-RI
20 μL Sterile
£71.86
Pack of 960
In Stock
to see stock.
Edge
Add to basket
11535504
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
3731-05-RI
300 μL Sterile
£67.20
Pack of 960
In Stock
to see stock.
Add to basket
11515504
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
3751-05-RI
200 μL Sterile
£66.95
Pack of 960
Estimated Shipment: 11-07-2025
to see stock.
Add to basket
11555504
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
3781-05-RI
1000 μL Sterile
£60.45
Pack of 768
In Stock
to see stock.
Edge
Add to basket
Description
Description
Pipette samples with the accuracy and precision you expect from your pipette every time, with Thermo Scientific™ SoftFit-L™ pipette tips. Low retention tips deliver maximum volume of sample and are for use with Rainin™ LTS pipettes.
  • Reload towers are an environmentally-friendly packaging option that reduces plastic waste and storage space
  • Compatible with SoftFit-L empty racks
  • Color-coded inserts allows quick identification of tip volume
Specifications
Specifications
Non-filtered
DNase, RNase, DNA, ATP, and Endotoxin Free
Universal
ART SoftFit L
Natural
Refill
Rainin LTS LiteTouch
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ SoftFit-L™ Non-Filtered Low Retention Pipette Tip in Reload Tower