Learn More
3M™ Molecular Detection Assay 2
Used with the 3M™ Molecular Detection System for the rapid and specific pathogen detection of E. coli O157, including H7, in enriched food samples.
Brand:  3M™ 7100089364
Encompass
Product Code. 17268540
  • £1001.18 / 1 set
Estimated Shipment: 21-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • One assay protocol and fewer steps than most tests reduces repetitive tasks and makes it easy to train employees.
  • Ready-to-use and pre-dispensed reagents provide convenience and minimize the chance of human error.
  • Color-coded assay tubes by organism and matching software screens allow for better tracking and easy identification.
  • The ability to run all assays simultaneously and the flexibility to test up to 96 samples in each run helps you operate more efficiently.
  • Real-time amplification and detection process identifies positive results in as early as 15 minutes and allows you to make critical decisions faster.
Specifications
Specifications
Escherichia coli
E. coli O157
Microbiology Media
96