missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 2-NBDG (2-(N-(7-Nitrobenz-2-oxa-1,3-diazol-4-yl)Amino)-2-Deoxyglucose)

Product Code. 11569116
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11569116

Brand: Invitrogen™ N13195

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

2-NBDG is a fluorescent glucose analog that has been used to monitor glucose uptake in live cells, as an indicator of cell viability.

2-NBDG is a fluorescent glucose analog that has been used to monitor glucose uptake in live cells, as an indicator of cell viability. Although sensitive to its environment NBD fluorescence typically displays excitation/emission maxima of ∼465/540 nm and can be visualized using optical filters designed for fluorescein.

Dissolve the dye in water or 1X PBS (pH 7.2) to prepare a stock solution up to 10 mg/mL. Stock solutions should be prepared fresh and used on same day.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5°C to -30°C) and protect from light.
Detection Method Fluorescence
For Use With (Equipment) Fluorescence Microscope, Flow Cytometer
Product Type 2-NBDG
Dye Type Other Label(s) or Dye(s)
Format Tube(s), Slide(s)
Quantity 5 mg
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.