Learn More
Invitrogen™ Alexa Fluor™ 594 Phalloidin
High-affinity F-actin probe conjugated to bright, photostable, red fluorescent Alexa Fluor 594 dye
Brand: Invitrogen™ A12381
581.65 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 50
Additional Details : Weight : 0.00454kg Transport : UN number : 2811 Chem class : 6.1 Pack group : II
Description
Get Superior Results in Actin Staining Studies
Phalloidin is a bicyclic peptide belonging to a family of toxins isolated from the deadly Amanita phalloides “death cap” mushroom and is commonly used in imaging applications to selectively label F-actin. Fluorescently-labeled phalloidin has virtually identical binding properties with actin from different species including plants and animals. Phalloidin binds F-actin with high selectivity while Alexa Fluor 594 provides red fluorescence of unparalleled brightness and photostability. Demonstrating very little nonspecific staining, Alexa Fluor 594 phalloidin allows high-contrast discrimination of actin staining.
- Selectively stains F-actin
- Outstanding fluorescence performance
- Excitation/Emission: 581/609nm
- Superior to antibody staining
- Optimal for fixed and permeabilized samples
Cell Analysis, Cell Structure, Cytoskeleton. Used to visualize and quantitate F-actin in tissue sections, cell cultures, or cell-free preparations
Order Info
Shipping Condition: Room Temperature
Specifications
Store in freezer -5°C to -30°C and protect from light. | |
Alexa Fluor™ 594 | |
Room Temperature | |
Phalloidin |
581⁄609 | |
Alexa Fluor™ | |
Alexa Fluor Dyes | |
Actin, Cytoskeleton |
Safety and Handling
- Alexa Fluor 594 phalloidin
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
Hazard Statement
- H300-Fatal if swallowed.
- H310-Fatal in contact with skin.
- H330-Fatal if inhaled.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P262-Do not get in eyes, on skin, or on clothing.
- P264-Wash hands thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P330-Rinse mouth.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ALEXA FLUOR 594 Phalloidin
For Research Use Only. Not for use in diagnostic procedures.