Learn More
Invitrogen™ Alexa Fluor™ 647-aha-dCTP
For better resolution of two-color microarray gene expression assays
Brand: Invitrogen™ A32771
Code : 50
Additional Details : Weight : 0.00450kg
Description
The Alexa Fluor™ 647 dyes, used to label nucleotides, are compatible with commonly used microarray scanners, and provide greater signal correlation (R2) valuesthan the spectrally similar Cy™3 and Cy5 dye pair, improving the resolution of two-color microarray gene expression assays to 1.3-fold changes in expression. The exceptionally bright and photostable Alexa Fluor dyes are also essentially insensitive to pH and are highly water soluble.
Array CGH, DNA and RNA Purification and Analysis, DNA Labeling, Gene Expression Analysis and Genotyping, Genotyping and Genomic Profiling, Nucleic Acid Labeling and Oligo Synthesis
Order Info
Shipping Condition: Wet Ice
Specifications
Labeled Nucleotide | |
Microarray Gene Expression Assay | |
Alexa Fluor Dye | |
AHA (5-aminohexylacrylamido) | |
Alexa Fluor™ |
1 mM | |
Store in freezer (-5 to -30°C) and protect from light. | |
Direct Labeling | |
Wet Ice | |
50 μL |
For Research Use Only. Not for use in diagnostic procedures.