Learn More
Invitrogen™ Albumin from Bovine Serum (BSA), Alexa Fluor™ 488 conjugate
Bright and photostable conjugate exceptionally suited for applications in which well-defined molecular weight tracers are needed
Brand: Invitrogen™ A13100
Code : 50
Additional Details : Weight : 0.00525kg
Description
- Applications include studying rate of endocytosis and exocytosis and following intracellular protein processing
Cell Analysis, Cell Tracing and Tracking, Cellular Imaging, General Cell Tracing, Immunofluorescence (IF), Immunofluorescence Staining and Detection
Order Info
Shipping Condition: Room temperature
Specifications
5 mg | |
Bovine | |
Alexa Fluor Dyes | |
Lyophilized | |
Alexa Fluor™ |
Alexa Fluor 488 | |
Store in freezer (-5 to -30°C) and protect from light. | |
Room Temperature | |
Albumin Proteins |
For Research Use Only. Not for use in diagnostic procedures.