missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Amplex™ Red Glucose/Glucose Oxidase Assay Kit

Format allows for multiple time point measurements

Brand:  Invitrogen™ A22189

374.64 GBP valid until 2024-11-29
Use promo code "21683" to get your promotional price.


Product Code. 10494392

  • £412.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

The Amplex Red Glucose/Glucose Oxidase Assay Kit provides a sensitive and simple method for detecting d-glucose or glucose oxidase using a fluorescence microplate reader or fluorometer. The Amplex Red reagent (10-acetyl-3,7-dihydroxyphenoxazine) is a colorless, stable, and extremely versatile peroxidase substrate. Because peroxidase- and glucose oxidase-mediated reactions can be coupled, it is possible to measure glucose oxidase activity or the release of glucose by any glucosidase enzyme—for instance, β-glucosidase and glucocerebrosidase—in either a continuous or discontinuous assay. Because of the ability to couple reactions, this assay has been demonstrated to be useful for the quantification of glucose levels in foods, fermentation media, and bodily fluids. In the assay, glucose oxidase reacts with d-glucose to form d-gluconolactone and hydrogen peroxide. In the presence of horseradish peroxidase (HRP), hydrogen peroxide then reacts with the Amplex Red reagent in a 1:1 stoichiometric ratio to generate the red-fluorescent product resorufin. Because resorufin has absorption and fluorescence emission maxima of approximately 571nm and 585nm, respectively, there is little interference from autofluorescence in most biological samples.

  • Detect concentrations as low as 3μM d-glucose or 0.05mU/mL of glucose oxidase
  • Format allows for multiple time point measurements
  • Designed for minimal autofluorescence interference
  • Custom assay design and packaging are also available

Use Amplex Red Assays for a Broad Range of Investigations
A wide variety of validated Amplex Red assays are available for studying cell signaling and lipids, neurobiology, inflammation and immune function, and metabolism. We also offer Amplex UltraRed Reagent, a second-generation reagent providing greater sensitivity and brighter fluorescence, and the Amplex Red/UltraRed Stop Reagent. The Amplex Red/UltraRed Stop Reagent provides convenience and control by allowing the fluorescence signal-generating reaction to be terminated at a user-determined time point. After addition of the stop reagent, the fluorescence signal remains stable for at least three hours.

Enzyme and Protein Activity Assays, Oxidase and Peroxidase Activity, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store in freezer -5°C to -30°C and protect from light.
Glucose/Glucose Oxidase Assay
Red Glucose/Glucose Oxidase Assay Kit
Tube(s), 96-well plate
500 assays
Fluorescence
Microplate Reader, Spectrophotometer, Fluorometer
Other Label(s) or Dye(s)
Amplex™
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.