Learn More
Invitrogen™ Amplex™ Red Hydrogen Peroxide/Peroxidase Assay Kit
Sensitive, one-step assay to detect hydrogen peroxide (H2O2) or peroxidase activity
Brand: Invitrogen™ A22188
379.05 GBP valid until 2025-12-31
Use promo code "24276" to get your promotional price.
Description
Assay uses AmplexRed reagent (10-acetyl-3,7-dihydroxyphenoxazine) to detect hydrogen peroxide (H2O2) or peroxidase activity in vitro
. AmplexRed reagent, in combination with horseradish peroxidase (HRP), can detect H2O2 released from biological samples, including cells, or generated in enzyme-coupled reactions. AmplexRed reagent can also be used as an ultrasensitive assay for peroxidase activity when H2O2 is in excess. In the presence of peroxidase, AmplexRed reagent reacts with H2O2 in a 1:1 stoichiometry to produce the red-fluorescent oxidation product, resorufin. Resorufin has excitation and emission maxima of approximately 571 nm and 585 nm, respectively, and because the extinction coefficient is high (58,000 ± 5000cm–1M–1), the assay can be performed fluorometrically or spectrophotometrically. This reaction has been used to detect as little as 10 picomoles of H2O2 in a 100μL volume or 1 x 10–5U/mL of HRP.
- Can detect as little as 10 picomoles of hydrogen peroxide or 10μU/mL of horseradish peroxidase activity in a 100μL assay volume
- Provides sufficient reagents for approximately 500 assays using reaction volumes of 100μL per assay
Cell Analysis, Cell Culture, Cell Viability, Proliferation and Function, Enzyme and Protein Activity Assays, Nitro-Oxidative Stress, Oxidase and Peroxidase Activity, Proteins, Expression, Isolation and Analysis, Reactive Oxygen Species, Stem Cell Analysis, Stem Cell Labeling and Detection, Stem Cell Research
Order Info
Shipping Condition: Room Temperature

Specifications
Store in freezer -5°C to -30°C and protect from light. | |
Hydrogen Peroxidase Assay | |
Red Hydrogen Peroxide/Peroxidase Assay Kit | |
Tube(s), 96-well plate | |
500 assays |
Fluorescence | |
Microplate Reader, Spectrophotometer, Fluorometer | |
Other Label(s) or Dye(s) | |
Amplex™ | |
Room Temperature |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.